ContactUsLinkComponent
Grating Replicas: Parallel and Crossed Lines, Electron Microscopy Sciences
Grating Replicas: Parallel and Crossed Lines, Electron Microscopy Sciences
  100501-382
Minority or Woman-Owned Business Enterprise
undefined
Grating Replicas: Parallel and Crossed Lines, Electron Microscopy Sciences
  100501-382
 :  80051
Minority or Woman-Owned Business Enterprise
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
  • • Issued document with your organization's Federal Tax ID Number
  • • Government issued document with your organization's Resale Tax ID Number
  • • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
  • Description:
    Grating Replica, Cross Line
  • Environmentally Preferable:
  • Cat. no.:
    100501-382
  • Supplier No.:
    80051
Parallel and crossed line grating replicas are calibration specimens for transmission electron microscopy and scanning transmission electron microscopy.
Parallel: Shadowcast carbon replicas of diffraction parallel line gratings with spacing of 462.9 nm, 2160 lines per millimeter onto 3.05 mm grid.
Crossed: Cross line pitch spacing of 500 nm with 2000 lines/mm in both directions, cross at 90° to one another give additional accuracy to magnification checks and aid in checking distortion. Trench-type groove makes it easy to measure pitch. Ideal for calibration electron-optical magnification up to the ×80000/100000 range.