Glass Scale, Electron Microscopy Sciences
Glass Scale, Electron Microscopy Sciences
Glass Scale, Electron Microscopy Sciences
Glass Scale, Electron Microscopy Sciences
A transparent glass plate well polished and marked by the photo-printing technique.
It provides very accurate readings and it is a helpful tool for the stereo viewer.
Product Family Options
  • Désignation
    Matière
    Graduation
    Longueur
    Width
    Épaisseur
  • Scale 200, marking area: 200 mm
    Glass
    0.1 mm
    220 mm
    25 mm
    3 mm
    Each
    Each - $902.78
    $902.78
    Each
       $902.78
     : 68142
  • Scale 50, marking area: 50 mm
    Glass
    0.1 mm
    80 mm
    25 mm
    3 mm
    Each
    Each - $346.99
    $346.99
    Each
       $346.99
     : 68140