Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
Glass Sample Bottles, With Plastic Snap Cap, Electron Microscopy Sciences
For collecting and exhibiting a wide variety of specimens in the lab or field. Caps snap over a special sealing ring to form an effective barrier to moisture, making these bottles especially suitable for hydroscopic materials.
Product Family Options
  • Capacité
    Désignation
    Taille
  • 32 ml
    With Plastic Snap Cap
    32 ml
    Each
    Each - $1,008.16
    $1,008.16
    Pack of 144
    Pack of 144 - $768.65
    $768.65
    Each
       $1,008.16
    Pack of 144
       $768.65
     : 64260
  • Replacement Snap Cap
    22 mm
    Each
    Each - $95.37
    $95.37
    Pack of 200
    Pack of 200 - $83.30
    $83.30
    Each
       $95.37
    Pack of 200
       $83.30
     : 64251-22
  • 4 ml
    With Plastic Snap Cap
    4 ml
    Each
    Each - $745.84
    $745.84
    Pack of 144
    Pack of 144 - $568.94
    $568.94
    Each
       $745.84
    Pack of 144
       $568.94
     : 64250
  • 8 ml
    With Plastic Snap Cap
    8 ml
    Each
    Each - $581.50
    $581.50
    Pack of 144
    Pack of 144 - $466.26
    $466.26
    Each
       $581.50
    Pack of 144
       $466.26
     : 64252
  • 12 ml
    With Plastic Snap Cap
    12 ml
    Each
    Each - $724.50
    $724.50
    Pack of 144
    Pack of 144 - $544.80
    $544.80
    Each
       $724.50
    Pack of 144
       $544.80
     : 64254
  • 16 ml
    With Plastic Snap Cap
    16 ml
    Each
    Each - $536.99
    $536.99
    Pack of 144
    Pack of 144 - $488.11
    $488.11
    Each
       $536.99
    Pack of 144
       $488.11
     : 64256
  • 24 ml
    With Plastic Snap Cap
    24 ml
     : -
  • Replacement Snap Cap
    30 mm
    Each
    Each - $138.47
    $138.47
    Pack of 200
    Pack of 200 - $104.66
    $104.66
    Each
       $138.47
    Pack of 200
       $104.66
     : 64257-30