Glass Microbeakers, Electron Microscopy Sciences
À propos de cet article
Used for the processing of micro-samples and for visual or microscopic examination, staining, fixation, infiltration, liquid evaporation, bacterial growth, weighing, etc.
3 Options Available Below
Product Details & Documents
Used for the processing of micro-samples and for visual or microscopic examination, staining, fixation, infiltration, liquid evaporation, bacterial growth, weighing, etc.
Product Family Options
Informations sur le produit
- CapacitéHauteurØ ext.AvailabilityPrice
- 1 ml (0.03 oz.)13 mm12 mmBox of 100$746.27Each$746.27Supplier Number: 60980
- 7.5 ml25 mm25 mmBox of 100$1,198.74Each$1,198.74Supplier Number: 60986
- 2.5 ml13 mm21 mmBox of 100$870.83Each$870.83Supplier Number: 60982