Glass Coplin Staining Jar, Screw Cap, Electron Microscopy Sciences | Avantor
Glass Coplin Staining Jar, Screw Cap, Electron Microscopy Sciences
5-Slide Unit Glass Coplin Jar, Screw Cap
Catalog # 100500-232
Fournisseur #:  70317
Glass Coplin Staining Jar, Screw Cap, Electron Microscopy Sciences
Catalog # 100500-232
Glass Coplin Staining Jar, Screw Cap, Electron Microscopy Sciences
Glass Coplin Staining Jar, Screw Cap, Electron Microscopy Sciences
5-Slide Unit Glass Coplin Jar, Screw Cap
Price
$476.59
Log in pour la disponibilité
Case of 4 Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Staining Jar holds 5 single 1 x 3" slides vertically, or 10 slides back-to-back.
Slides stay within the jar so that it can be capped during staining. Complete with a screw cap and a metal foil liner.
Overall measurement: 4 3/8 x 2 3/4 x 2 3/8.
Spécifications
  • Désignation:
    5-Slide Unit Glass Coplin Jar, Screw Cap