Foreline Traps, Electron Microscopy Sciences
Foreline Traps, Electron Microscopy Sciences
Foreline Traps, Electron Microscopy Sciences
Certificates
Foreline Traps, Electron Microscopy Sciences
Foreline Traps, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
These rechargeable in-line traps effectively block oil backstreaming from mechanical pumps. Any mechanically pumped system, even one with a turbo pump, should have a foreline traps. The trap is placed in the foreline between the roughing pump and the diffusion or turbo pump. It uses a disposable, oxygen-free copper maze absorbent which requires no liquid nitrogen and will not hold water. Typically the maze is replaced every 1-2 years. The seamless body is available in aluminum. Replacement elements (Copper Maze) also available.
  • A quick change clamp for replacement of maze
  • A viton “O” ring end cap
  • A recessed screen
  • Available in five pipe diameters
Product Family Options
Product Information
  • Désignation
    Capacité
    Matière
    Availability
    Price
  • Foreline Traps 3/4"
    5 CFM
    Aluminum
    Log in for availability
    Log in for availability
    Each
    Each - $1,045.26
    $1,045.26
    Each
       $1,045.26
    Supplier Number: 91050-15
  • Foreline Traps 11/2"
    10 CFM
    Aluminum
    Log in for availability
    Log in for availability
    Each
    Each - $1,045.26
    $1,045.26
    Each
       $1,045.26
    Supplier Number: 91050-25
  • Foreline Traps 1"
    10 CFM
    Aluminum
    Log in for availability
    Log in for availability
    Each
    Each - $1,045.26
    $1,045.26
    Each
       $1,045.26
    Supplier Number: 91050-20