Folding Magnifier, Electron Microscopy Sciences
Lightweight, compact, and simple; it comes with a measuring scale on the base frame.
Base size is 1 square inch opening; marked every 1/8" on two sides and every millimeter on two other sides. Comes with 6X magnifier lens in a black anodized aluminum body. Folds flat when it is not in use.
- Grossissement:6×
- Désignation:Folding Magnifier