About this item
The 6" Wooden Shaft Applicator is round, foam-tip over regular-cotton tip on a 6" long wooden shaft
These foam tipped applicators are lint free, non-abrasive, and sized for precision cleaning with solvents. (Freon, Trichloroethane, Isopropyl Alcohol, and Acetone). The head is made from 100% urethane foam with 100 PPI porosity. Tips are shred resistant and bonded securely to the shaft.
The 6" Wooden Shaft Applicator is round, foam-tip over regular-cotton tip on a 6" long wooden shaft. Head size d x L: 3/8 x 1 3/16". Packed 50 sticks in a sealed bag.
The 5" Shaft, Rectangular Foam Tip is polypropylene shaft. Head size d x L: 1 1/16 x 1". Packed 50 sticks in a sealed bag.
The 6" Wooden Shaft Anti-Static Foam-Tip Over Cotton Tip measures d x L: 3/8 x 1 3/16". Packed 50 sticks in a sealed bag.
Head size d x L: 3/8 x 1 3/16". Packed 50 sticks in a sealed bag.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.