To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
- Description:Fluoromount G
- Size:25 ml
- Cat. no.:CA100502-406
A special mounting media for immunofluorescent staining procedures. It is a water soluble, non-fluorescing medium which is well suited for use when the staining procedure has an aqueous final step. It contains 10% polyvinyl alcohol in phosphate buffered glycerol with 0.1% sodium azide as a preservative.
A special mounting media for immunofluorescent staining procedures. It is a water soluble, non-fluorescing medium which is well suited for use when the staining procedure has an aqueous final step. It contains 10% polyvinyl alcohol in phosphate buffered glycerol with 0.1% sodium azide as a preservative.
In light microscopy, tissues were stained with conventional histological stains (e.g.,hematoxylin-eosin) and embedded in Micro-Bed: the results were comparable with those provided on paraffin embedded tissues. Further, polychrome silver-based and cytochemical stains (e.g.,PAS) were also feasible. In transmission electron microscopy, images as good as those obtained with similar acrylic resins, (e.g., Lowicryl K4M), were obtained.