Flexible Shaft Machine - General Application Kit, Electron Microscopy Sciences
A versatile hang-up flexible shaft machine.
Accessories information: Mounted accessories are on 2.35 mm (3/32") arbors.
Ordering information: Available either 1/10 or 1/8 HP ball bearing motor, variable speed 0 to 18 rpm. Complete kit comes with a hand piece, 1 m (39") Flexible shaft, variable speed foot pedal, and a bracket for hanging. Lifetime warranty on motor under normal use.
A versatile hang-up flexible shaft machine.
Accessories information: Mounted accessories are on 2.35 mm (3/32") arbors.
Ordering information: Available either 1/10 or 1/8 HP ball bearing motor, variable speed 0 to 18 rpm. Complete kit comes with a hand piece, 1 m (39") Flexible shaft, variable speed foot pedal, and a bracket for hanging. Lifetime warranty on motor under normal use.
Product Family Options
Flexible Shaft Machine, 110 V
- DésignationTension
- : 62130-10
- 1/10 Hp Flex Shaft Machine Set110 V-: 62131-10
- : 62133-10
- : 62134-10
Flexible Shaft Machine, 220 V
- DésignationTension
- 1/8 Hp Flex Shaft Machine220 V-: 62130-20
- 1/10 Hp Flex Shaft Machine Set220 V-: 62131-20
- : 62133-20
- : 62134-20