Five-Slide Gripper, Electron Microscopy Sciences | Avantor
Five-Slide Gripper, Electron Microscopy Sciences
Five Slide Gripper
Catalog # 100499-574
Fournisseur #:  71410-06
Five-Slide Gripper, Electron Microscopy Sciences
Catalog # 100499-574
Five-Slide Gripper, Electron Microscopy Sciences
Five-Slide Gripper, Electron Microscopy Sciences
Five Slide Gripper
Price
$172.87
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Five-slide gripper accomodates microscope slides in one staining procedure. It is made from a special material which is resistant to all chemicals and solvents which are used in staining.
  • No need to remove slides for drying
  • Drying temps up to 80 °C
  • Fits coplin or round-open staining jars
Spécifications
  • Désignation:
    Five Slide Gripper