Finger Cots, Latex, Electron Microscopy Sciences
À propos de cet article
Inexpensive, expandable finger cots.
- Seamless
- Protective dexterity without covering the entire hand
- Powderless
- Non-rolled
3 1/2 mil (0.0030"). Length 40 mm (2 3/4").
3 Options Available Below
Product Details & Documents
Inexpensive, expandable finger cots.
- Seamless
- Protective dexterity without covering the entire hand
- Powderless
- Non-rolled
3 1/2 mil (0.0030"). Length 40 mm (2 3/4").
Product Family Options
Informations sur le produit
- TailleAvailabilityPrice
- LargeEach29,26 $Pack of 14422,39 $Supplier Number: 71132
- MediumEach25,69 $Pack of 14419,67 $Supplier Number: 71131
- SmallEach24,41 $Pack of 14418,69 $Supplier Number: 71130