Fairs NG5 Graticule, Electron Microscopy Sciences | Avantor
Fairs NG5 Graticule, Electron Microscopy Sciences
Fairs NG5 Graticule, Electron Microscopy Sciences
Certificates
Fairs NG5 Graticule, Electron Microscopy Sciences
Fairs NG5 Graticule, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Designed to extend the sizing range of globe and circle graticules. Example: Used in conjunction with NG2 the overall size range =128:1. The circles increase by 2.
Caution: Note: Both graticules would have to be used with the same microscope, eyepiece and objective.
Product Family Options
Informations sur le produit
  • Taille
    Availability
    Price
  • 16 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $702.66
    $702.66
    Each
       $702.66
    Supplier Number: 68033-16
  • 19 mm
    Discontinued
    Supplier Number: 68033-19
  • 21 mm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $702.66
    $702.66
    Each
       $702.66
    Supplier Number: 68033-21