Tissue Chopper, Electron Microscopy Sciences
Tissue Chopper, Electron Microscopy Sciences
Tissue Chopper, Electron Microscopy Sciences
Certificates
Tissue Chopper, Electron Microscopy Sciences
Tissue Chopper, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
The TC-1 Tissue Chopper is designed to facilitate the cutting of 100 to 1000 micron thick brain tissue slices for use in recording experiments.
The chopper includes a guillotine arm with adjustments to assure blade alignment. The brain sample is moved in respect to the arm with a stage whose position can be read with 2 to 3 micron precision on a digital micrometre scale. The stage is made with a removable "PTFE" surface plate to facilitate cleaning.
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • TC-1 Tissue Chopper with Micrometer
    Log in for availability
    Log in for availability
    Each
    Each - $9,250.96
    $9,250.96
    Each
       $9,250.96
    Supplier Number: 93102
  • Replacement PFTE Plate
    Log in for availability
    Log in for availability
    Each
    Each - $259.66
    $259.66
    Each
       $259.66
    Supplier Number: 93110