About this item
Fabricated from genuine Lucite®; these racks fit into our 5×7" developing tanks. One end of the rack is extended in length with a cut-out hole to serve as a handle.
The racks are designed to accommodate a number of sheets per rack without changing the length of the rack. As the number of the films per rack increases, the space between them decreases.
Some Products May Appear Restricted
To ensure a smooth and speedy checkout, please log in to your account. Some items may show as restricted simply because you're not logged in.
If you do not have an account, you can register using our registration webform (https://www.avantorsciences.com/ca/en/login/register)
If you're still seeing restrictions after logging in, certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order. Some items may additionally require a specific license or customer documentation; additional documentation will be requested for these items prior to shipment.