EMS Diamond Knife Wash, Electron Microscopy Sciences
Product Details & Documents
This new concept of washing the Diamond knives using magnetic stirring action (with special stirring bar) gives you a fast, clean and safe method to remove particles that are left on the knives edge after sectioning.
Since most of the EM labs already have stirring plates, the Diamond Knife Wash has been designed for use with your existing unit, saving you the need to purchase an extra one. It is universal and can be used for all type and brands of Diamond Knives, including the Diatome Histo Knife.
Warning: Never use on a hot plate. If used on a combination Stirrer/Hot Plate, make sure the heat is off.
Specifications
- Désignation:Diamond Knife Wash