Embedding Rings, Electron Microscopy Sciences
Embedding Rings, Electron Microscopy Sciences
Embedding Rings, Electron Microscopy Sciences
Embedding Rings, Electron Microscopy Sciences
Molded from the highest quality plastic, our Embedding Rings fit most base molds. For added convenience, there are four colors from which to choose Blue, Pink, Green, Yellow. The large writing surface further eases identification and handling. The base measures 1 5/8 x 1 1/8" and the ring measures 25 x 25 mm (1 x 1").
Product Family Options
  • Désignation
    Color
  • Embedding ring
    Yellow
    Box of 250
    Box of 250 - $119.94
    $119.94
    Each
    Each - $119.94
    $119.94
    Box of 250
       $119.94
    Each
       $119.94
     : 62350-Y
  • Embedding ring
    Green
    Box of 250
    Box of 250 - $119.94
    $119.94
    Each
    Each - $119.94
    $119.94
    Box of 250
       $119.94
    Each
       $119.94
     : 62350-G
  • Embedding ring
    White
    Box of 250
    Box of 250 - $136.31
    $136.31
    Each
    Each - $136.31
    $136.31
    Box of 250
       $136.31
    Each
       $136.31
     : 62350-W
  • Embedding Ring, Pink
    Box of 250
    Box of 250 - $113.31
    $113.31
    Each
    Each - $113.31
    $113.31
    Box of 250
       $113.31
    Each
       $113.31
     : 62350-P