Embedding Capsule; Flat, Electron Microscopy Sciences | Avantor
Embedding Capsule; Flat, Electron Microscopy Sciences
Flat Tip Embedding Capsules, 39O.D. x 6dia. mm (19/16 x 1/4")
Catalog # 100488-336
Fournisseur #:  70021
Embedding Capsule; Flat, Electron Microscopy Sciences
Catalog # 100488-336
Embedding Capsule; Flat, Electron Microscopy Sciences
Embedding Capsule; Flat, Electron Microscopy Sciences
Flat Tip Embedding Capsules, 39O.D. x 6dia. mm (19/16 x 1/4")
Price
$224.81
Log in pour la disponibilité
Each Pack of 40
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Added to our extensive line of embedding capsules is our flat bottom capsules, which are made from polyethylene, and are 8 mm (just like the standard 00 capsules) in diameter.
Spécifications
  • Désignation:
    Flat Tip Embedding Capsules
  • Dimensions:
    39O.D. x 6dia. mm (19/16 x 1/4")