Embedding Blocks, Electron Microscopy Sciences
About this item
Embedding blocks are precisely machined from hard phenolic resin
It is suitable for cellodin or paraffin sections. The faces of the embedding blocks are cut very deeply providing a firm hold of the embedded specimen.
4 Options Available Below
Product Details & Documents
Embedding blocks are precisely machined from hard phenolic resin
It is suitable for cellodin or paraffin sections. The faces of the embedding blocks are cut very deeply providing a firm hold of the embedded specimen.
Product Family Options
Product Information
- DésignationDimensionsAvailabilityPrice
- Supplier Number: 62363-01
- Supplier Number: 62363-02
- Supplier Number: 62363-03
- Supplier Number: 62363-04