Ellipso-Spoon® Sampler Spoons, Electron Microscopy Sciences
About this item
The Ellipso-Spoon® Sampler has a spoon end and a spatula end and is made from polished stainless steel.
Its unique shape means no more material loss. This utensil’s specially curved shape permits contact with all corners of bottles and containers; therefore, no loss of material occurs. Crystals, powders, granulated and solid substances, and even thick liquids are easily retrieved and never fall back into the container.
3 Options Available Below
Product Details & Documents
The Ellipso-Spoon® Sampler has a spoon end and a spatula end and is made from polished stainless steel.
Its unique shape means no more material loss. This utensil’s specially curved shape permits contact with all corners of bottles and containers; therefore, no loss of material occurs. Crystals, powders, granulated and solid substances, and even thick liquids are easily retrieved and never fall back into the container.
Product Family Options
Product Information