Edwards RV8 Vacuum Pump, Electron Microscopy Sciences
À propos de cet article
Rotary vacuum pump; Double stage. Ultra Grade 19 oil is recommended for use with the RV8. Weight 50 lbs (24 kg).
- 6.9 cfm (195 L/min) displacement
- Ultimate vacuum (without gas ballast) 1.5x10-3 torr
- Max. inlet water pressure vapor: 29 torr
- Max. water vapor pumping rate: .48 lb/h
- Motor: 3/4 hp
- Oil capacity Max/Min 0.75L/0.43
1 Options Available Below
Product Details & Documents
Rotary vacuum pump; Double stage. Ultra Grade 19 oil is recommended for use with the RV8. Weight 50 lbs (24 kg).
- 6.9 cfm (195 L/min) displacement
- Ultimate vacuum (without gas ballast) 1.5x10-3 torr
- Max. inlet water pressure vapor: 29 torr
- Max. water vapor pumping rate: .48 lb/h
- Motor: 3/4 hp
- Oil capacity Max/Min 0.75L/0.43
Product Family Options
Informations sur le produit
- DésignationElectricalAvailabilityPrice
- Supplier Number: 91025-E
- RV8 Vacuum Pump110 VDiscontinued-Supplier Number: 91025