Duplex Specimen, Electron Microscopy Sciences | Avantor
Duplex Specimen, Electron Microscopy Sciences
Duplex Reference Specimen
Catalog # 100494-988
Fournisseur #:  79521-01
Duplex Specimen, Electron Microscopy Sciences
Catalog # 100494-988
Duplex Specimen, Electron Microscopy Sciences
Duplex Specimen, Electron Microscopy Sciences
Duplex Reference Specimen
Price
$2,755.90
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
An alternative and very sensitive test is by means of an alloy with two major Copper/Zinc phases separated by an atomic number difference of 0.1. The light phase illustrated in the micrograph has a mean atomic number of 29.47 and the dark phase a mean atomic number of 29.37.
Spécifications
  • Désignation:
    Duplex Reference Specimen