Double Neubauer Ruled Hemacytometer, Electron Microscopy Sciences
À propos de cet article
The complete set includes one counting chamber, two 0.5 mm cover glasses, one red cell pipette, one white blood cell pipette, tubing, mouthpieces and carrying case.
An “H” shape moat which forms 2 counting areas are metal plated and double Neubauer ruled providing sharp bright line contrast for consistent cell counts. A “V” cut on each area is used to control the possibility of overflow. Triple dividing lines clearly show cells within the counting area. The ruled surface is 0.1 mm below the cover glass; liquid capacity is 0.1 mm3/mm2 and 0.00025 mm3/grid square.
The complete set includes one counting chamber, two 0.5 mm cover glasses, one red cell pipette, one white blood cell pipette, tubing, mouthpieces and carrying case.
3 Options Available Below
Product Details & Documents
The complete set includes one counting chamber, two 0.5 mm cover glasses, one red cell pipette, one white blood cell pipette, tubing, mouthpieces and carrying case.
An “H” shape moat which forms 2 counting areas are metal plated and double Neubauer ruled providing sharp bright line contrast for consistent cell counts. A “V” cut on each area is used to control the possibility of overflow. Triple dividing lines clearly show cells within the counting area. The ruled surface is 0.1 mm below the cover glass; liquid capacity is 0.1 mm3/mm2 and 0.00025 mm3/grid square.
The complete set includes one counting chamber, two 0.5 mm cover glasses, one red cell pipette, one white blood cell pipette, tubing, mouthpieces and carrying case.
Product Family Options
Informations sur le produit
- DésignationAvailabilityPrice
- Supplier Number: 63510-10
- Phase Counting Chamber (3200)Discontinued-Supplier Number: 63510-20
- Supplier Number: 63510-13
- Supplier Number: 63510-11