Dissecting Scissors - Operating Scissors, Electron Microscopy Sciences
Dissecting Scissors - Operating Scissors, Electron Microscopy Sciences
Dissecting Scissors - Operating Scissors, Electron Microscopy Sciences
Dissecting Scissors - Operating Scissors, Electron Microscopy Sciences
They come in a variety of styles and lengths:
Made from stainless steel.
They come in a variety of styles and lengths:
S/B - One sharp point and one blunt point
S/S - Two sharp points
B/B - Two blunt points
Product Family Options
  • Désignation
    Longueur
    Blade Type
  • Curved
    5½"
    Sharp/Blunt
    Each
    Each - $31.07
    $31.07
    Each
       $31.07
     : 72966-01
  • Curved
    4½"
    Sharp/Blunt
    Each
    Each - $23.71
    $23.71
    Each
       $23.71
     : 72964-02
  • Straight
    5½"
    Sharp/Sharp
    Each
    Each - $39.46
    $39.46
    Each
       $39.46
     : 72966-02
  • Straight
    5½"
    Blunt/Blunt
    Each
    Each - $37.32
    $37.32
    Each
       $37.32
     : 72966-04
  • Curved
    6½"
    Sharp/Blunt
    Each
    Each - $37.32
    $37.32
    Each
       $37.32
     : 72968-02
  • Curved
    4½"
    Blunt/Blunt
    Each
    Each - $23.71
    $23.71
    Each
       $23.71
     : 72964-03
  • Straight
    6½"
    Blunt/Blunt
    Each
    Each - $43.21
    $43.21
    Each
       $43.21
     : 72968-05
  • Straight
    6½"
    Sharp/Sharp
    Each
    Each - $43.88
    $43.88
    Each
       $43.88
     : 72968-03
  • Straight
    4½"
    Blunt/Blunt
    Each
    Each - $25.74
    $25.74
    Each
       $25.74
     : 72964-01
  • Curved
    5½"
    Blunt/Blunt
     : 72966-05
  • Curved
    5½"
    Sharp/Sharp
    Each
    Each - $35.77
    $35.77
    Each
       $35.77
     : 72966-03
  • Curved
    6½"
    Blunt/Blunt
     : 72968-06
  • Straight
    6½"
    Sharp/Blunt
    Each
    Each - $43.21
    $43.21
    Each
       $43.21
     : 72968-01
  • Curved
    6¼"
    Sharp/Sharp
    Each
    Each - $43.21
    $43.21
    Each
       $43.21
     : 72968-04