Dissecting Dish, Electron Microscopy Sciences
À propos de cet article
A silicone pad placed inside a 150x20 mm deep plastic petri dish. The mat can be removed, washed, and returned to the dish for reuse. The pins come in 10 different coded colors.
Optional Pyrex Glass Petri dish is available for use when sterilization is needed.
2 Options Available Below
Product Details & Documents
A silicone pad placed inside a 150x20 mm deep plastic petri dish. The mat can be removed, washed, and returned to the dish for reuse. The pins come in 10 different coded colors.
Optional Pyrex Glass Petri dish is available for use when sterilization is needed.