Dip Miser, For Slide Coating, 10 ml Capacity, Electron Microscopy Sciences
Supplier: Electron Microscopy Sciences
Certificates
About this item
Slide coating cup with stand and cleaning brush. Glass cup is Pyrex and is designed to conserve expensive autoradiographic emulsion when coating glass slides. Needs only 1/4 the volume of solution previously required. Single piece holder permits immersion in a water bath and provides a stable darkroom stand to prevent spills, and faster temperature adjustment of the emulsion.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products, please provide identification that includes your business name and shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of identification are:
- • Issued document with your organization's Federal Tax ID Number
- • Government issued document with your organization's Resale Tax ID Number
- • Any other Government ID that includes the business name and address
Avantor will not lift restrictions for residential shipping addresses.
Frequently Bought Together