Diamond Tip Scribers, Electron Microscopy Sciences
About this item
Diamond tip scribers are in wide use throughout the laboratory. It leaves a permanent marking on virtually any surface, including metal, glass, or plastic.
- Hexagonal aluminum pencil shaped body with a mill finish
- Natural diamond tip
- Protective cap included
- Straight and Angle tip scribers are available
4 Options Available Below
Product Details & Documents
Diamond tip scribers are in wide use throughout the laboratory. It leaves a permanent marking on virtually any surface, including metal, glass, or plastic.
- Hexagonal aluminum pencil shaped body with a mill finish
- Natural diamond tip
- Protective cap included
- Straight and Angle tip scribers are available
Product Family Options
Product Information
- DésignationDiameterLongueurPoint SizeAvailabilityPrice
- Diamond Tip Scriber, 60°, Straight Head1/4" (handle)6"1/8"Box of 6$268.38Each$268.38Supplier Number: 70031
- Supplier Number: 70032
- Diamond Scriber, 75° Angle Tip1/8" (handle)6½"1/16" Ø x 1/4" LEach$288.86Pack of 6$199.98Supplier Number: 70033
- Supplier Number: 70036
Accessories
Product Information
- Catalog #AvailabilityPrice
- Refill for Diamond Tip Retractable Scriber, 60° Tip FinishRefill for Diamond Tip Retractable Scriber, 60° Tip Finish100491-584Log in for availabilityEach - $43.13