Diamond Stone, Electron Microscopy Sciences | Avantor
Diamond Stone, Electron Microscopy Sciences
Diamond Stone, All Purpose Fine Grit
Catalog # 100501-578
Fournisseur #:  62120
Diamond Stone, Electron Microscopy Sciences
Catalog # 100501-578
Diamond Stone, Electron Microscopy Sciences
Diamond Stone, Electron Microscopy Sciences
Diamond Stone, All Purpose Fine Grit
Price
$31.88
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This diamond stone has millions of cutting edges to sharpen most any tool.
Faster than dry or oil stones
Used as a file or a stone
Cuts in all directions
Available in all purpose fine grit
Spécifications
  • Désignation:
    Diamond Stone, All Purpose Fine Grit