Diamond Points, Electron Microscopy Sciences | Avantor
Diamond Points, Electron Microscopy Sciences
Diamond Points, Electron Microscopy Sciences
Certificates
Diamond Points, Electron Microscopy Sciences
Diamond Points, Electron Microscopy Sciences
5 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
All diamond points are mounted on 2.35 mm (3/32”) shanks. Suitable for Flex Shaft machines. Exceptionally long lasting. Use at 10,000 RPM.
Product Family Options
Informations sur le produit
  • Désignation
    Ø
    Shape
    Availability
    Price
  • Diamond Points, Inverted
    Cone
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $59.89
    $59.89
    Each
       $59.89
    Supplier Number: 62155-23
  • Diamond Points, Square Grid
    3.8 mm
    Cone
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $65.77
    $65.77
    Each
       $65.77
    Supplier Number: 62155-38
  • Diamond Points, Small
    1.3 mm
    Round
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $53.41
    $53.41
    Each
       $53.41
    Supplier Number: 62155-13
  • Diamond Points, Flat & Cylinder
    3.5 mm
    Flat/Cylinder
    Discontinued
    Supplier Number: 62155-35
  • Diamond Points, Square Edge
    6.3 mm
    Square
    Discontinued
    Supplier Number: 62155-63
  • Diamond Points, Inverted Wide
    8.3 mm
    Cone
    Discontinued
    Supplier Number: 62155-83
  • Diamond Points, Large
    2.9 mm
    Round
    Discontinued
    Supplier Number: 62155-29
  • Diamond Points, Tapered
    7.3 mm
    Wheel
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $65.77
    $65.77
    Each
       $65.77
    Supplier Number: 62155-73
  • Diamond Points, Cone
    2.4 mm
    Cone
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $57.07
    $57.07
    Each
       $57.07
    Supplier Number: 62155-24