Diamond Lapping Film, Electron Microscopy Sciences
Diamond Lapping Film, Electron Microscopy Sciences
Certificates
Diamond Lapping Film, Electron Microscopy Sciences
Diamond Lapping Film, Electron Microscopy Sciences
7 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A selective diamond particle resin bonded to a uniform 3-mil polyester film backing. Excellent for polishing samples with differing hardness. Ideal for the preparation of cross sections for SEM/TEM analysis. Comes in 8" and 12" discs with plain back (P/B) or pressure sensitive adhesive back (PSA).
Product Family Options
Informations sur le produit
  • Granulométrie
    Backing
    Taille
    Availability
    Price
  • 0.25 µm
    Plain back
    8"
    Discontinued
    Supplier Number: 50350-03
  • 3 µm
    PSA
    12"
    Discontinued
    Supplier Number: 50355-15
  • 0.25 µm
    Plain back
    12"
    Discontinued
    Supplier Number: 50354-03
  • 0.25 µm
    PSA
    12"
    Discontinued
    Supplier Number: 50355-03
  • 9 µm
    PSA
    12"
    Discontinued
    Supplier Number: 50355-25
  • 15 µm
    PSA
    12"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $219.71
    $219.71
    Each
       $219.71
    Supplier Number: 50355-30
  • 0.1 µm
    PSA
    8"
    Discontinued
    Supplier Number: 50351-01
  • 6 µm
    Plain back
    8"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $91.41
    $91.41
    Each
       $91.41
    Supplier Number: 50350-20
  • 15 µm
    Plain back
    8"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $96.05
    $96.05
    Each
       $96.05
    Supplier Number: 50350-30
  • 0.5 µm
    PSA
    8"
    Discontinued
    Supplier Number: 50351-05
  • 1 µm
    PSA
    8"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $97.69
    $97.69
    Each
       $97.69
    Supplier Number: 50351-10
  • 0.5 µm
    Plain back
    12"
    Discontinued
    Supplier Number: 50354-05
  • 1 µm
    Plain back
    12"
    Discontinued
    Supplier Number: 50354-10
  • 30 µm
    Plain back
    12"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $219.73
    $219.73
    Each
       $219.73
    Supplier Number: 50354-35
  • 30 µm
    PSA
    12"
    Discontinued
    Supplier Number: 50355-35
  • 0.25 µm
    PSA
    8"
    Discontinued
    Supplier Number: 50351-03
  • 9 µm
    Plain back
    8"
    Discontinued
    Supplier Number: 50350-25
  • 9 µm
    PSA
    8"
    Discontinued
    Supplier Number: 50351-25
  • 1 µm
    Plain back
    8"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $90.15
    $90.15
    Each
       $90.15
    Supplier Number: 50350-10
  • 15 µm
    PSA
    8"
    Discontinued
    Supplier Number: 50351-30
  • 3 µm
    Plain back
    12"
    Discontinued
    Supplier Number: 50354-15
  • 0.5 µm
    PSA
    12"
    Discontinued
    Supplier Number: 50355-05
  • 0.1 µm
    Plain back
    8"
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $96.40
    $96.40
    Each
       $96.40
    Supplier Number: 50350-01
  • 30 µm
    PSA
    8"
    Discontinued
    Supplier Number: 50351-35
  • 9 µm
    Plain back
    12"
    Discontinued
    Supplier Number: 50354-25