Desiccant Canister; Reusable, Electron Microscopy Sciences | Avantor
Desiccant Canister; Reusable, Electron Microscopy Sciences
Desiccant Canister; Reusable, Electron Microscopy Sciences
Certificates
Desiccant Canister; Reusable, Electron Microscopy Sciences
Desiccant Canister; Reusable, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Ideal for dry control of desiccator cabinets. Moisture is absorbed through the perforations in the aluminum case, which contains silica gel. This 2 x 4" canister maintains a safe level of humidity in 3 cu.ft (0.8 cu. meter) area. When the monitor window turns green the silica gel is saturated. It can be regenerated in a 300 °F (150 °C) oven in about 3 hours. The monitor window will be pink in color when recharged.
Product Family Options
Informations sur le produit
  • Désignation
    Availability
    Price
  • Desiccant Canister
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $322.69
    $322.69
    Log in pour la disponibilité
    Pack of 4
    Pack of 4 - $240.73
    $240.73
    Each
       $322.69
    Pack of 4
       $240.73
    Supplier Number: 71211
  • Desiccant Canister
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $82.79
    $82.79
    Each
       $82.79
    Supplier Number: 71210