Darkroom Thermometer, Electron Microscopy Sciences
Darkroom Thermometer, Electron Microscopy Sciences
Darkroom Thermometer, Electron Microscopy Sciences
Darkroom Thermometer, Electron Microscopy Sciences
Model A comes with 1 3/4" dial with a 6" stem.
Precision Dial Thermometer: easy to read Fahrenheit and Centigrade scales. Hermetically sealed; water proof; will not fog; made from corrosion resistant stainless steel. Comes with an adjustable positioning clamp. Accuracy to 1/2°.
Model A comes with 1 3/4" dial with a 6" stem.
Model B comes with 2 1/2" luminescent dial with a 8" stem, 1/2° graduations and a calibration adjustment wrench.
Product Family Options
  • Désignation
  • Darkroom Thermometer, Model A
    Each
    Each - $86.36
    $86.36
    Each
       $86.36
     : 74388
  • Luminescent Thermometer, Model B
    Each
    Each - $114.60
    $114.60
    Each
       $114.60
     : 74389