Crystalbond™, Electron Microscopy Sciences | Avantor
Crystalbond™, Electron Microscopy Sciences
Crystalbond™, Electron Microscopy Sciences
Certificates
Crystalbond™, Electron Microscopy Sciences
Crystalbond™, Electron Microscopy Sciences
3 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Temporary adhesives that can be washed away after machining.
  • Machining or slicing single crystal metal specimens
  • Grinding and polishing sapphire, ceramic, optical garnets, ferrites, and LCD glass
  • Dicing and slicing germanium and silicon wafers in semiconductor production
  • Dicing and slicing alumina and beryllia substrates for IC and microelectronic production
  • Holding beam leads in IC devices for pull-off tests
  • Dicing subminiature chip capacitors and microwave IC substrates
These wash away temporary adhesives are used as a temporary bond for holding delicate crystals, metallurgical specimens, glass components, and ceramic substrates for dicing, slicing, drilling and polishing
Crystalbond™ 509:
Offers excellent adhesion to metals, glass, and ceramic. This material is the best for precise high purity work since it leaves no residue after dissolving and does not clog the diamond wheel as compared to conventional waxes. Each stick weighs about 90 grams.
Crystalbond™ 555:
The same as 509 but used in applications where it is desirable to have a temporary bond which is water soluble. Each stick weighs about 90 grams.
Crystalbond™ 590:
The same as 509 but offers maximum adhesion to subminiature parts. Each stick weighs about 225 grams.
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • Crystalbond 555
    90 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $639.45
    $639.45
    Log in pour la disponibilité
    Pack of 10
    Pack of 10 - $536.79
    $536.79
    Each
       $639.45
    Pack of 10
       $536.79
    Supplier Number: 50401-10
  • Crystalbond 590, 1 Stick
    225 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $189.86
    $189.86
    Each
       $189.86
    Supplier Number: 50402-01
  • Crystalbond 509
    90 g
    Discontinued
    Supplier Number: 50400-05
  • Crystal Bond Stripper-590, 100g
    100 g
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $92.63
    $92.63
    Each
       $92.63
    Supplier Number: 50402-S