Cryo-Lok Cryogenic Vials, Electron Microscopy Sciences
Cryo-Lok Cryogenic Vials, Electron Microscopy Sciences
Certificates
Cryo-Lok Cryogenic Vials, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
CYO-LOK vials are ideal for storage or transport of material of extremely low temperatures.
  • Graduated in 0.5 ml increments
The vials have external threads, providing a smooth and uniform inner surface. This eliminates the problem of air born contamination and/or sample loss so common when vials with internal threads are used. The screw top incorporates a molded-in sealing ring, which assures a tight leak-proof seal.
Product Family Options
Informations sur le produit
  • Capacité
    Avec bouchon
    Sterility
    Availability
    Price
  • 2.50 ml
    Oui
    Non sterile
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Box of 1,000
    Box of 1,000 - $258.43
    $258.43
    Log in pour la disponibilité
    Each
    Each - $258.43
    $258.43
    Box of 1,000
       $258.43
    Each
       $258.43
    Supplier Number: 72652
  • Discontinued
    Supplier Number: -
  • Discontinued
    Supplier Number: -
  • 2.50 ml
    Oui
    Sterile
    Discontinued
    Supplier Number: -
  • Discontinued
    Supplier Number: -