Cotton Tipped Applicators, Electron Microscopy Sciences | Avantor
Cotton Tipped Applicators, Electron Microscopy Sciences
Cotton Tipped Applicators, Electron Microscopy Sciences
Certificates
Cotton Tipped Applicators, Electron Microscopy Sciences
Cotton Tipped Applicators, Electron Microscopy Sciences
4 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Lint-free, cotton-tipped applicators are suitable for cleaning delicate parts. Measures 6" in length, 1/12" Ø wooden shaft and head size is 3/16" (Ø) x 5/8" (L).
Handy Vial Packs: 3" (8 cm) wooden shaft, small-tipped applicators come pre-packed in a plastic-capped vial, a handy way to keep a supply of swabs available anytime on the bench top.
Product Family Options
Informations sur le produit
  • Désignation
    Sterility
    Packaging
    Availability
    Price
  • Cotton Tipped Applicator, Single End
    Non sterile
    100/Pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 6,14 $
    6,14 $
    Log in pour la disponibilité
    Pack of 100
    Pack of 100 - 6,11 $
    6,11 $
    Each
       6,14 $
    Pack of 100
       6,11 $
    Supplier Number: 72310
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100489-878
    Minority or Woman-Owned Business Enterprise
  • Cotton Tipped Applicator, Single End
    Non sterile
    10 Pack/Case
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 72,02 $
    72,02 $
    Each
       72,02 $
    Supplier Number: 72310-10
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100499-648
    Minority or Woman-Owned Business Enterprise
  • Cotton-Tipped Applicator
    Non sterile
    1000/Pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 399,01 $
    399,01 $
    Each
       399,01 $
    Supplier Number: 72310-S
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100493-712
    Minority or Woman-Owned Business Enterprise
  • Handy Vial Pack
    Sterile
    10 Vials/Pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - 94,20 $
    94,20 $
    Each
       94,20 $
    Supplier Number: 72307
    Spécifications:
    Minority or Woman-Owned Business Enterprise
    More
    Specifications
    Cat. No.
    100496-340
    Minority or Woman-Owned Business Enterprise