Cryo Labels, Electron Microscopy Sciences
Cryo Labels, Electron Microscopy Sciences
Cryo Labels, Electron Microscopy Sciences
Certificates
Cryo Labels, Electron Microscopy Sciences
Cryo Labels, Electron Microscopy Sciences
1 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A special formulated material for use at ultra-low temperatures and in cryo storage conditions, such as liquid nitrogen and oxygen.
  • Will not peel or shrink
They are durable and adhere well to plastic as well as cardboard cryogenic storage boxes. Ten sheets of labels: five preprinted, five plain.
Packaging: Packaged as 20 labels/sheet.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    Packaging
    Availability
    Price
  • Cryo labels
    25×50 mm
    200 labels/pack
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $253.40
    $253.40
    Log in pour la disponibilité
    Pack of 200
    Pack of 200 - $229.89
    $229.89
    Each
       $253.40
    Pack of 200
       $229.89
    Supplier Number: 62046
  • Cryo labels
    25×50 mm
    10 packs/lot
    Discontinued
    Supplier Number: 62047