Cryo-Gloves®, Electron Microscopy Sciences
À propos de cet article
Now offer cryogenic personal protective garments of the highest quality.
- Features protection from –135 to 150 °C
- Superior thermal protection and water-resistant material
- Is comfortable, lightweight and has excellent dexterity
- Breathable with stay-dry lining
Cryo-Gloves® and Cryo-Aprons® are essential accessories for those working in ultra cold environments. Providing effective protection in a wide variety of applications. Freezers are ultra low temperature for dry ice handling. Used for liquid nitrogen, liquid oxygen and liquid helium.
Best in clinical laboratories, Bio-Medical environments, blood banks, frozen food processing labs and pharmaceutical labs.
15 Options Available Below
Product Details & Documents
Now offer cryogenic personal protective garments of the highest quality.
- Features protection from –135 to 150 °C
- Superior thermal protection and water-resistant material
- Is comfortable, lightweight and has excellent dexterity
- Breathable with stay-dry lining
Cryo-Gloves® and Cryo-Aprons® are essential accessories for those working in ultra cold environments. Providing effective protection in a wide variety of applications. Freezers are ultra low temperature for dry ice handling. Used for liquid nitrogen, liquid oxygen and liquid helium.
Best in clinical laboratories, Bio-Medical environments, blood banks, frozen food processing labs and pharmaceutical labs.
Specifications for Product Family
Spécifications
- Catalog No:
- 102091-168
- 102091-170
- 102091-172
- 102091-174
- 102091-176
- 102091-178
- 102091-180
- 102091-182
- 102091-184
- 102097-308
- 102097-310
- 102097-312
- 102097-314
- 102097-316
- 102097-318
- 102097-320
- Ambidextrous/Hand-specific:
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
- Ambidextrous
Spécifications
Product Family Options
Informations sur le produit
- DésignationTaille du gantGlove ColorLongueurAvailabilityPrice
- WristSBlue30.5 cm (12")Each (1Pair)$441.45Pair$452.04Supplier Number: 71070Spécifications:
More
Specifications
Cat. No.102091-168Ambidextrous/Hand-specificAmbidextrous - WristMBlue30.5 cm (12")Each (1Pair)$441.45Pair$452.04Supplier Number: 71071Spécifications:
More
Specifications
Cat. No.102091-170Ambidextrous/Hand-specificAmbidextrous - WristLBlue30.5 cm (12")Each (1Pair)$441.45Pair$452.04Supplier Number: 71072Spécifications:
More
Specifications
Cat. No.102091-172Ambidextrous/Hand-specificAmbidextrous - Mid ArmSBlue35.6 cm (14")Each (1Pair)$494.32Pair$506.18Supplier Number: 71073Spécifications:
More
Specifications
Cat. No.102091-174Ambidextrous/Hand-specificAmbidextrous - Mid ArmMBlue35.6 cm (14")Each (1Pair)$494.32Pair$506.18Supplier Number: 71074Spécifications:
More
Specifications
Cat. No.102091-176Ambidextrous/Hand-specificAmbidextrous - Mid ArmLBlue35.6 cm (14")Each (1Pair)$494.38Pair$506.25Supplier Number: 71075Spécifications:
More
Specifications
Cat. No.102091-178Ambidextrous/Hand-specificAmbidextrous - ElbowSBlue45.7 cm (17⅞")Each (1Pair)$574.30Pair$588.08Supplier Number: 71076Spécifications:
More
Specifications
Cat. No.102091-180Ambidextrous/Hand-specificAmbidextrous - ElbowLBlue45.7 cm (17⅞")Each (1Pair)$574.30Pair$588.08Supplier Number: 71077Spécifications:
More
Specifications
Cat. No.102091-182Ambidextrous/Hand-specificAmbidextrous - ElbowXLBlue45.7 cm (17⅞")Each (1Pair)$574.30Pair$588.08Supplier Number: 71078Spécifications:
More
Specifications
Cat. No.102091-184Ambidextrous/Hand-specificAmbidextrous - WristXLBlue30.5 cm (12")Each (1Pair)$441.45Pair$452.04Supplier Number: 71072-XSpécifications:
More
Specifications
Cat. No.102097-308Ambidextrous/Hand-specificAmbidextrous - Mid ArmXLBlue35.6 cm (14")Each (1Pair)$494.32Pair$506.18Supplier Number: 71075-XSpécifications:
More
Specifications
Cat. No.102097-310Ambidextrous/Hand-specificAmbidextrous - ElbowMBlue45.7 cm (17⅞")Each (1Pair)$574.30Pair$588.08Supplier Number: 71076-MSpécifications:
More
Specifications
Cat. No.102097-312Ambidextrous/Hand-specificAmbidextrous - ShoulderLBlue68.5 - 71.1 cm (26⅞ - 27⅞")Discontinued-Supplier Number: 71079-LSpécifications:
More
Specifications
Cat. No.102097-314Ambidextrous/Hand-specificAmbidextrous - ShoulderMBlue68.5 - 71.1 cm (26⅞ - 27⅞")Each (1Pair)$795.77Pair$814.87Supplier Number: 71079-MSpécifications:
More
Specifications
Cat. No.102097-316Ambidextrous/Hand-specificAmbidextrous - Shoulder Cryo Glove, SmallSBlue68.5 - 71.1 cm (26⅞ - 27⅞")Each (1Pair)$795.77Pair$814.87Supplier Number: 71079-SSpécifications:
More
Specifications
Cat. No.102097-318Ambidextrous/Hand-specificAmbidextrous - Shoulder Cryo Glove, Extra LargeXLBlue68.5 - 71.1 cm (26⅞ - 27⅞")Each (1Pair)$795.77Pair$814.87Supplier Number: 71079-XLSpécifications:
More
Specifications
Cat. No.102097-320Ambidextrous/Hand-specificAmbidextrous