CoverWell™ Imaging Chambers, Electron Microscopy Sciences
CoverWell™ Imaging chambers are designed to stabilize and support thick and free-floating specimens for confocal microscopy and imaging applications.
CoverWell™ Imaging chambers are designed to stabilize and support thick and free-floating specimens for confocal microscopy and imaging applications.
Product Family Options
- DésignationTailleProfondeur
- 1 well, 50 µl20 mm dia2.5 mm-: 70327-25
- 1 well, 180 µl20 mm dia0.8 mmEach$495.37Pack of 40$365.64: 70327-08
- 1 well, 180 µl20 mm dia0.6 mmEach$495.80Pack of 40$377.97: 70327-05
- 1 well, 200 µl28 mm dia0.26 mmEach$450.65Pack of 25$325.07: 70327-04
- 1 well, 300 µl20 mm dia1.3 mmEach$502.62Pack of 20$344.69: 70327-13
- 1 well, 300 µl20 mm dia0.9 mmEach$477.95Pack of 40$364.35: 70327-10
- 1 well, 520 µl20 mm dia1.7 mmEach$284.30Pack of 20$206.92: 70327-20
- 1 well, 750 µl20 mm dia2.8 mm-: 70327-28