Coverslip Holders, Electron Microscopy Sciences
Coverslip Holders, Electron Microscopy Sciences
Coverslip Holders, Electron Microscopy Sciences
Certificates
Coverslip Holders, Electron Microscopy Sciences
Coverslip Holders, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Coverslips can be stacked with spacers (up to 15 coverslips with a thickness up to two mm) with samples placed on top of each coverslip.
  • Brass/Nickel plated
  • Fluids freely exchange between coverslips
  • Both spacers and weights come with the unit
The upper part of the holder is grooved to accept tweezers.
Product Family Options
Informations sur le produit
  • Désignation
    Dimensions
    No. of Places
    Availability
    Price
  • Coverslip Holder 1 for CPD, 9 - 13 mm Ø coverslips
    16×24 mm
    10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $978.51
    $978.51
    Each
       $978.51
    Supplier Number: 70193-01
  • Coverslip Holder 2 for CPD, 16 - 22 mm Ø coverslips
    24×25 mm
    10
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $984.14
    $984.14
    Each
       $984.14
    Supplier Number: 70193-02