Coverglass Staining Rack, Electron Microscopy Sciences | Avantor
Coverglass Staining Rack, Electron Microscopy Sciences
Coverglass Staining Rack, 12.7 mm (5")
Catalog # 100491-942
Fournisseur #:  72240
Coverglass Staining Rack, Electron Microscopy Sciences
Catalog # 100491-942
Coverglass Staining Rack, Electron Microscopy Sciences
Coverglass Staining Rack, Electron Microscopy Sciences
Coverglass Staining Rack, 12.7 mm (5")
Price
566,49 $
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Coverglass staining rack is made from polished stainless steel.
  • Holds five of 22×24 mm coverglasses
  • Comes with a handle, 12.7 mm (5") long
This rack is specially designed to be used with coplin jars.
Spécifications
  • Désignation:
    Coverglass Staining Rack
  • Longueur:
    12.7 mm (5")