COVAMAT® Lining, Electron Microscopy Sciences
About this item
Versatile clear lining material: clear polyethylene ribbed matting can be used for lining laboratory bench top, shelves, trays, and more.
It is easily trimmed and also works well under drums or carboys. 0.25 mm (0.010") width 0.7 mm (0.030") thick ribs, 76 cm (30") wide, 15.2 m (50') and 4.6 m (15') roll. 1 roll/box.
1 Options Available Below
Product Details & Documents
Versatile clear lining material: clear polyethylene ribbed matting can be used for lining laboratory bench top, shelves, trays, and more.
It is easily trimmed and also works well under drums or carboys. 0.25 mm (0.010") width 0.7 mm (0.030") thick ribs, 76 cm (30") wide, 15.2 m (50') and 4.6 m (15') roll. 1 roll/box.
Product Family Options
Product Information
- DésignationCapacitéAvailabilityPrice
- Covamat Lining, 15'15 ftDiscontinued-Supplier Number: -
- Covamat Lining, 50'50 ftEach (50Feet)$528.87Roll of 50$528.87Supplier Number: 72615-50