Cotton Tipped Applicator, Electron Microscopy Sciences
À propos de cet article
A lint-free, 6" double cotton tipped applicator with one pointed end and one regular end on a 1/12" wooden shaft.
The pointed head size is 3/16" tapering to 1/32 x 5/8". Regular head size 3/16 x 5/8".
Packaging: Packed 100 sticks in a sealed bag.
4 Options Available Below
Product Details & Documents
A lint-free, 6" double cotton tipped applicator with one pointed end and one regular end on a 1/12" wooden shaft.
The pointed head size is 3/16" tapering to 1/32 x 5/8". Regular head size 3/16 x 5/8".
Packaging: Packed 100 sticks in a sealed bag.
Product Family Options
Informations sur le produit
- DésignationHandle LengthAvailabilityPrice
- Double End, Tapered, Wooden Shaft16 cm (6")Each$30.58Pack of 100$23.40Supplier Number: 72308
- Supplier Number: 72308-05
- Large Tip, Wooden Shaft16 cm (6")Each$40.90Pack of 5$31.76Supplier Number: 72309
- Supplier Number: 72306