Compartment - Micro Processing/Embedding Cassettes, Electron Microscopy Sciences
À propos de cet article
Made from Acetal Polymer. These biopsy cassettes can hold up to six tissue specimens, each one placed in it’s own compartment, numbered from 1 to 6 and measuring 7 x 12 x 5 mm (1/4 x 7/16 x 3/16"). Cover and base have over 3000 0.36 mm square openings. Cassettes keep specimen safely submerged in solvent and are totally resistant to the most histological solvents. The efficient flow-through slots maximize fluid exchange and insure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. Large labeling areas are located on three sides of the cassette. The anterior writing area is at a 45° angle, making the cassette, to some degree, able to be used with certain types of cassette labeling instruments. Each case contains 4 dispenser boxes of 250 cassettes with covers assembled.
9 Options Available Below
Product Details & Documents
Made from Acetal Polymer. These biopsy cassettes can hold up to six tissue specimens, each one placed in it’s own compartment, numbered from 1 to 6 and measuring 7 x 12 x 5 mm (1/4 x 7/16 x 3/16"). Cover and base have over 3000 0.36 mm square openings. Cassettes keep specimen safely submerged in solvent and are totally resistant to the most histological solvents. The efficient flow-through slots maximize fluid exchange and insure proper drainage. The one-piece integral lid eliminates the need for separate steel lids. Large labeling areas are located on three sides of the cassette. The anterior writing area is at a 45° angle, making the cassette, to some degree, able to be used with certain types of cassette labeling instruments. Each case contains 4 dispenser boxes of 250 cassettes with covers assembled.
Product Family Options
Informations sur le produit
- ColorAvailabilityPrice
- LilacBox of 250$481.17Each$481.17Supplier Number: 70078-L
- PinkBox of 500$558.57Each$558.57Supplier Number: 70078-P
- BlueBox of 500$481.17Each$481.17Supplier Number: 70078-B
- GrayBox of 250$481.17Each$481.17Supplier Number: 70078-GR
- AquaBox of 250$566.50Each$566.50Supplier Number: 70078-A
- YellowBox of 500$481.17Each$481.17Supplier Number: 70078-Y
- PeachDiscontinued-Supplier Number: 70078-PE
- OrangeBox of 250$558.57Each$558.57Supplier Number: 70078-O
- TanBox of 250$468.81Each$468.81Supplier Number: 70078-T
- GreenBox of 500$566.50Each$566.50Supplier Number: 70078-G