Compact Slide Box for 25 Slides, Electron Microscopy Sciences
Compact Slide Box for 25 Slides, Electron Microscopy Sciences
Compact Slide Box for 25 Slides, Electron Microscopy Sciences
Certificates
Compact Slide Box for 25 Slides, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Made from durable polystyrene (with heavy walls).
Same characteristics as the Secured Hinge Microscope Slide Boxes, but with a slip-on cover. Holds 25 1x3" slides. Measures H x W x L: 1/4 x 3 1/2 x 5 1/2".
Product Family Options
Product Information
  • Désignation
    Availability
    Price
  • 25 Capacity
    Log in for availability
    Log in for availability
    Log in for availability
    Bag of 2
    Bag of 2 - $47.34
    $47.34
    Log in for availability
    Each
    Each - $47.34
    $47.34
    Bag of 2
       $47.34
    Each
       $47.34
    Supplier Number: 71460
  • 25 Capacity
    Log in for availability
    Log in for availability
    Each
    Each - $324.57
    $324.57
    Each
       $324.57
    Supplier Number: 71461