Colormark ™; Plus Glass Slide, Electron Microscopy Sciences
À propos de cet article
Specimen slide identification is vital automated etched slide labeling instruments ensure specimen slide integrity but are difficult to read on ordinary slides.
To clearly read diamond scribed slide labels against any surface, you need the only slide designed to be used with your automated slide labeling instrument.
You need COLORMARK slides.These brightly printed slides greatly improve etched labeling visibility and offer all the advantages of Superfrost slides. A bright contrasting color has been printed on the back of the Superfrost end so that patient or specimen information etched through the painted end on the front is clearly visible. Colormark Plus slides are adhesive coated therefore ensuring the adhesion of the section to the slide during staining procedures.
7 Options Available Below
Product Details & Documents
Specimen slide identification is vital automated etched slide labeling instruments ensure specimen slide integrity but are difficult to read on ordinary slides.
To clearly read diamond scribed slide labels against any surface, you need the only slide designed to be used with your automated slide labeling instrument.
You need COLORMARK slides.These brightly printed slides greatly improve etched labeling visibility and offer all the advantages of Superfrost slides. A bright contrasting color has been printed on the back of the Superfrost end so that patient or specimen information etched through the painted end on the front is clearly visible. Colormark Plus slides are adhesive coated therefore ensuring the adhesion of the section to the slide during staining procedures.
Product Family Options
Informations sur le produit
- DésignationColorCapacitéAvailabilityPrice
- Colormark slides, colored endsGreen Front & Back1 gDiscontinued-Supplier Number: 63400-30
- Supplier Number: 63400-41
- Colormark slides, colored endsLavender Front/Black Back1 gDiscontinued-Supplier Number: 63400-61
- Colormark slides, colored endsBlue Front & Back1 gEach$257.98Gross of 144$257.98Supplier Number: 63400-20
- Colormark slides, colored endsPink Front/Red Back1 gEach$298.06Gross of 144$298.06Supplier Number: 63400-40
- Colormark slides, colored endsWhite Front/Black Back1 gEach$282.97Gross of 144$282.97Supplier Number: 63400-10
- Colormark slides, colored endsYellow End Front/Red Back1 gEach$283.72Gross of 144$283.72Supplier Number: 63400-50
- Supplier Number: 63400-31
- Colormark slides, colored endsLavender Front/Black Back1 gEach$283.72Gross of 144$283.72Supplier Number: 63400-60