Colored Microscope Slide Boxes, Electron Microscopy Sciences
Colored Microscope Slide Boxes, Electron Microscopy Sciences
Colored Microscope Slide Boxes, Electron Microscopy Sciences
Certificates
Colored Microscope Slide Boxes, Electron Microscopy Sciences
Colored Microscope Slide Boxes, Electron Microscopy Sciences
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Molded from one piece (lid and bottom) polypropylene these slide boxes are able to accommodate either 25 or 100 standard 75 x 25 mm (3 x 1") slides.
The attached lid is held securely by a closed latch lock. Stacking feet maximize the stability when boxes are stacked up one atop the other. The boxes are unlined and the index number is printed on the bottom. Comes complete with a numbered slide index card. These boxes are available in blue, red or white.
Product Family Options
Informations sur le produit
  • Désignation
    Color
    Availability
    Price
  • 25 Capacity
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $27.89
    $27.89
    Each
       $27.89
    Supplier Number: 71458-W
  • 100 Capacity
    Red
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $31.41
    $31.41
    Each
       $31.41
    Supplier Number: 71459-R
  • 25 Capacity
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $27.54
    $27.54
    Each
       $27.54
    Supplier Number: 71458-B
  • 25 Capacity
    Red
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $27.54
    $27.54
    Each
       $27.54
    Supplier Number: 71458-R
  • 100 Capacity
    Blue
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $31.41
    $31.41
    Each
       $31.41
    Supplier Number: 71459-B
  • 100 Capacity
    White
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $31.41
    $31.41
    Each
       $31.41
    Supplier Number: 71459-W