Clip-on Magnifiers, Electron Microscopy Sciences
About this item
Clips on to any eyeglass frame leaving hands free.
Optical quality plastic lenses enables you to see tiny details without distortion using both eyes. Lightweight, comfortable and adjustable. Comes with carrying pouch.
1 Options Available Below
Product Details & Documents
Clips on to any eyeglass frame leaving hands free.
Optical quality plastic lenses enables you to see tiny details without distortion using both eyes. Lightweight, comfortable and adjustable. Comes with carrying pouch.
Product Family Options
Product Information
- DésignationGrossissementDioptersWorking DistanceAvailabilityPrice
- Supplier Number: 68260-07
- Clip On Magnifier1.5×2508 mm (20")Discontinued-Supplier Number: 68260-02