Chang Monolayer Molds, Electron Microscopy Sciences | Avantor
Chang Monolayer Molds, Electron Microscopy Sciences
Chang Monolayer Molds, Electron Microscopy Sciences
Certificates
Chang Monolayer Molds, Electron Microscopy Sciences
Chang Monolayer Molds, Electron Microscopy Sciences
2 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
This specially designed mold has proven useful in the processing of frozen sections, ascite cells, fresh imprints, as well as blood and marrow smears.
Designed by Dr. J.P. Chang for the embedding of a monolayer of cells.J. of Ultrastructural Res., 37, 370 (1971).
Product Family Options
Informations sur le produit
  • Désignation
    Taille
    Availability
    Price
  • 10 Cavities
    2 cmx2 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $38.85
    $38.85
    Each
       $38.85
    Supplier Number: 70920
  • 12 Cavities
    2 cm x 1 cm
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $39.97
    $39.97
    Each
       $39.97
    Supplier Number: 70910