Cartilage Knife, Electron Microscopy Sciences | Avantor
Cartilage Knife, Electron Microscopy Sciences
Cartilage Knife
Catalog # 100492-072
Fournisseur #:  72990
Cartilage Knife, Electron Microscopy Sciences
Catalog # 100492-072
Cartilage Knife, Electron Microscopy Sciences
Cartilage Knife, Electron Microscopy Sciences
Cartilage Knife
Price
$30.90
Log in pour la disponibilité
Each
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
A heavy duty stainless steel blade with a riveted wooden handle.
Blade length: 90 mm (3½"). Total length: 216 mm (8 1/2").
Spécifications
  • Désignation:
    Cartilage Knife