Carbon Rods, Electron Microscopy Sciences | Avantor
Carbon Rods, Electron Microscopy Sciences
Carbon Rods, Electron Microscopy Sciences
Certificates
Carbon Rods, Electron Microscopy Sciences
Carbon Rods, Electron Microscopy Sciences
6 Options Available Below
Some Products May Be Restricted
Certain products—like chemicals or medical devices—require additional account verification steps to be able to place an order or specific license or customer documentation. More info
Product Details & Documents
Carbon /graphite films are ideal in EM due to uniform amorphous form and high transparency to electron beams. Carbon/graphite rods are CVP™ (Chemical Vapor Process) purity and spectrographic grade, grade 1, produced to ASTM tolerances. Spectro-Grade rods are ideal for all TEM, SEM and all other critical applications.
  • Spectro-Grade graphite rods have an impurity level of 2 ppm or less with a maximum level element of each of 1 ppm
  • Technical grade carbon has a typical impurity level of 50 ppm or greater
  • For non critical SEM coating applications, technical grade may be used
Graphite is the stable form of carbon. Graphite is one of the softest and diamond is one of the hardest minerals known to man. Carbon is harder than graphite. For TEM and SEM applications, graphite is recommended.
Carbon /graphite films are most usable in TEM and SEM due to its value characteristics: uniformity amorphous and high transparent to electron beams. Because of low molecular weight and their unique structure (graphite have a sheet like structure, where the atoms all lie in a plane), carbon/graphite films have excellent mechanical stability, and even film thickness is about 1 – 2 nm.
Product Family Options
Informations sur le produit
  • Longueur
    Ø
    Désignation
    Availability
    Price
  • 304.8 mm (12")
    6.4 mm (1/4")
    Carbon Rods
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $328.42
    $328.42
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $250.32
    $250.32
    Each
       $328.42
    Pack of 12
       $250.32
    Supplier Number: 70212
  • 304.8 mm (12")
    3.2 mm (1/8")
    Carbon Rods
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $324.68
    $324.68
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $247.48
    $247.48
    Each
       $324.68
    Pack of 12
       $247.48
    Supplier Number: 70210
  • 304.8 mm (12")
    6.4 mm (1/4")
    Carbon (Graphite) Rods Spectro-Grade
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $433.20
    $433.20
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $330.17
    $330.17
    Each
       $433.20
    Pack of 12
       $330.17
    Supplier Number: 70230
  • 304.8 mm (12")
    3.2 mm (1/8")
    Carbon (Graphite) Rods Spectro-Grade
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $320.06
    $320.06
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $243.51
    $243.51
    Each
       $320.06
    Pack of 12
       $243.51
    Supplier Number: 70200
  • 304.8 mm (12")
    4.7 mm (316")
    Carbon (Graphite) Rods Spectro-Grade
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $224.17
    $224.17
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $171.43
    $171.43
    Each
       $224.17
    Pack of 12
       $171.43
    Supplier Number: 70231
  • 304.8 mm (12")
    3.2 mm (1/8")
    Carbon Rods Technical Grade
    Log in pour la disponibilité
    Log in pour la disponibilité
    Log in pour la disponibilité
    Each
    Each - $292.54
    $292.54
    Log in pour la disponibilité
    Pack of 12
    Pack of 12 - $221.72
    $221.72
    Each
       $292.54
    Pack of 12
       $221.72
    Supplier Number: 70301